主要文件
B2529
Sigma-Aldrich
半干转印仪
Complete unit
登录查看公司和协议定价
About This Item
UNSPSC代码:
41100000
NACRES:
NC.01
存货情况
not available in EU
正在寻找类似产品? 访问 产品对比指南
一般描述
极为适合于Western、Northern或Southern转膜,单元可接受最大20 x 20 cm并具有可变厚度的一块或多块胶。
不锈钢阴极和铂阳极板可产生高度均匀的电场并耐腐蚀。 Mylar护罩可遮盖阴极的未使用区域,防止产生电弧,确保所有电流流过凝胶。
Complete with two Mylar shields, safety-tip power leads and interlocking safety cover.
历史批次信息供参考:
分析证书(COA)
访问文档库
Naresh K Rajendran et al.
Oxidative medicine and cellular longevity, 2021, 6664627-6664627 (2021-01-29)
Increasing evidence suggests that adipose-derived stem cells (ADSCs) serve as a therapeutic approach for wound healing. The aim of this study was to determine the effect of photobiomodulation (PBM) on antioxidant enzymes in ADSCs. Four ADSC cell models, namely, normal