主要文件
Z219673
Sigma-Aldrich
Aldrich® Abderhalden drying apparatus
pkg of 1 ea
登录查看公司和协议定价
About This Item
UNSPSC代码:
41121800
NACRES:
NB.43
价格与库存信息目前不能提供
包装
pkg of 1 ea
相关类别
一般描述
The Aldrich Abderhalden drying apparatus has an improved design features a desiccant flask with a removable drying chamber. Multiple drying chambers may be used to dry several samples in series without dismantling and cleaning the main chamber, flask, and condenser assembly. Dried samples, sealed in the drying chamber, may be easily transferred to a dry box for sample removal under inert atmosphere.
Complete unit consists of a drying chamber (25.5 cm L × 25.4 mm I.D.), desiccant flask with a 2 mm glass stopcock, vapor chamber, Allihn condenser, and 250 mL boiling flask.
法律信息
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
历史批次信息供参考:
分析证书(COA)