主要文件
ZIQ7MSKT1
Milli-Q
Multi System Installation Kit for Milli-Q® IQ/IX/EQ Systems
Hydraulically connects up to 3 water purification systems to the same storage tank.
登录查看公司和协议定价
选择尺寸
1 KIT
$420.00
$420.00
请联系客服了解存货情况
索取更多信息
选择尺寸
变更视图
1 KIT
$420.00
About This Item
UNSPSC代码:
41104214
NACRES:
JA.13
$420.00
请联系客服了解存货情况
索取更多信息
特点
Hydraulic connection up to three purification system units to the same Milli-Q® IQ/IX storage tank
包装
pkg of 1 kit
相容性
for use with Milli-Q® EQ 7000
for use with Milli-Q® EQ 7008
for use with Milli-Q® EQ 7016
for use with Milli-Q® IX 7005
for use with Milli-Q® IX 7010
for use with Milli-Q® IX 7015
for use with Milli-Q® IQ 7003
for use with Milli-Q® IQ 7005
for use with Milli-Q® IQ 7010
for use with Milli-Q® IQ 7015
一般描述
This multi-system installation kit hydraulically connects up to 3 water purification units to the same Milli-Q® IQ/IX/EQ storage tank. For example, the kit allows you to connect two Milli-Q® IQ 7000 ultrapure water systems to a Milli-Q® IQ/IX/EQ tank that is being fed by a Milli-Q® IQ/IX 7003/05/10/15 system.
法律信息
Milli-Q is a registered trademark of Merck KGaA, Darmstadt, Germany