主要文件
ZFCBEQXAU
Milli-Q
Country box Milli-Q® EQ 7008/16 AU
Available only in Austria
登录查看公司和协议定价
About This Item
UNSPSC代码:
41104200
NACRES:
JA.13
价格与库存信息目前不能提供
相容性
for use with Milli-Q® EQ 7008
for use with Milli-Q® EQ 7016
一般描述
The Country box for the Milli-Q® EQ 7008/16 system is an installation accessory that contains a printed quick reference guide, a user manual in local language and in English on a USB key, and a power cord adapted to the local electrical network.
应用
The components of the Country box assist users to quickly access the information they need to correctly use their system and connect to the local power network.
法律信息
Milli-Q is a registered trademark of Merck KGaA, Darmstadt, Germany