主要文件
20-410
Sigma-Aldrich
Chromatin Assembly (Buffers) Kit
登录查看公司和协议定价
About This Item
UNSPSC代码:
41116158
NACRES:
NA.25
价格与库存信息目前不能提供
访问文档库
  1. Which document(s) contains shelf-life or expiration date information for a given product?
    If available for a given product, the recommended re-test date or the expiration date can be found on the Certificate of Analysis.
  2. How do I get lot-specific information or a Certificate of Analysis?
    The lot specific COA document can be found by entering the lot number above under the "Documents" section.
  3. Do I need Product No. 20410, Silanized Glass Wool for my packed column application?
    Most applications, except for nonreactive compounds such as hydrocarbons, PAHs, etc., need to be done using silanized glass wool.  If untreated glass wool is used and the compounds are reactive, such as basic compounds, the compound adsorbs to active silanol sites. This can result in peak tailing, loss of sensitivity on the low end and poor linearity.  For free acids, use acid-treated glass wool.   
  4. How do I find price and availability?
    There are several ways to find pricing and availability for our products. Once you log onto our website, you will find the price and availability displayed on the product detail page. You can contact any of our Customer Sales and Service offices to receive a quote.  USA customers:  1-800-325-3010 or view local office numbers.
  5. What is the Department of Transportation shipping information for this product?
    Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product. 
  6. My question is not addressed here, how can I contact Technical Service for assistance?
    Ask a Scientist here.