Merck
Z261394
Stainless steel 316 syringe needle, pipetting blunt 90° tip
gauge 14, L 4 in.
登录查看公司和协议定价
选择尺寸
1 EA
$169.00
$169.00
请联系客服了解存货情况
选择尺寸
变更视图
1 EA
$169.00
About This Item
UNSPSC代码:
41122002
eCl@ss:
32070500
NACRES:
NB.03
$169.00
请联系客服了解存货情况
制造商/商品名称
Popper 9870
长度
4 in.
计量器
14
正在寻找类似产品? 访问 产品对比指南
一般描述
For those applications requiring the use of plastic syringes with pipetting needles, these needles are specifically designed for secure, leakproof mating with plastic Luer tip disposable syringes and metal Luer tip glass syringes. Female Luer hubs and needles are made of 316 SS. Noncontaminating, easy to clean and resistant to corrosion.
历史批次信息供参考:
分析证书(COA)