主要文件
CAD9880
Dissolution cannulae
gauge 14, 316 stainless steel hub, L 4 3/8 in.
登录查看公司和协议定价
选择尺寸
1 EA
$1,460.00
$1,460.00
请联系客服了解存货情况
选择尺寸
变更视图
1 EA
$1,460.00
About This Item
UNSPSC代码:
41122002
NACRES:
NB.03
$1,460.00
请联系客服了解存货情况
物料
316 stainless steel hub
包装
pack of 1 ea
制造商/商品名称
Cadence Science 9880
长度
4 3/8 in.
计量器
14
正在寻找类似产品? 访问 产品对比指南
一般描述
Ideal for drug dissolution testing. Drug dissolution testing is routinely used in the pharmaceutical industry to provide critical release information for quality control purposes, in new drug development, and to predict in vivo drug release profiles. These cannula feature 316 stainless steel hub and tubing for maximum resistance to chemical reaction.
历史批次信息供参考:
分析证书(COA)