主要文件
Z243892
Sigma-Aldrich
Aldrich® splash protector
female joint: ST/NS 24/40, male joint: ST/NS 24/40
登录查看公司和协议定价
选择尺寸
1 EA
$202.00
$202.00
请联系客服了解存货情况
选择尺寸
变更视图
1 EA
$202.00
About This Item
UNSPSC代码:
41121815
NACRES:
NB.35
$202.00
请联系客服了解存货情况
接合处
female joint: ST/NS24/40
male joint: ST/NS24/40
正在寻找类似产品? 访问 产品对比指南
一般描述
Effectively prevents upward movement of foaming or bumping liquids in rotary evaporator traps. Unique design has two baffles with horizontally opposing holes that may also be used with other distillation techniques.
法律信息
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
历史批次信息供参考:
分析证书(COA)