主要文件
Z173258
Sigma-Aldrich
Reaction and storage flask, Kjeldahl, with glass stopcock on side-arm
capacity 50 mL, female joint: ST/NS 14/20
登录查看公司和协议定价
About This Item
UNSPSC代码:
41121804
NACRES:
NB.43
物料
borosilicate glass
round bottom
two-neck
标签
R
接合处
female joint: ST/NS14/20
无菌性
non-sterile
特点
closure type female Joint: ST/NS
包装
pack of 1
制造商/商品名称
Sigma-Aldrich
容量
50 mL
正在寻找类似产品? 访问 产品对比指南
相关类别
历史批次信息供参考:
分析证书(COA)
访问文档库
商品
Versatile system for chemical reactions, extractions, and transfers of solids and liquids in laboratories.