Z173126
1 of 10
Sigma-Aldrich
Z409235
Aldrich® Schlaker reaction tube
Z515981
Schlenk reaction and storage tube
Z166898
Aldrich® 气敏化学玻璃实验室套件
Z220434
Sealing tube
SYNF891410
Synthware™ reaction tube with glass stopcock
Z409294
Z173231
Reaction and storage flask, Kjeldahl, with glass stopcock on side-arm
Z173223
Reaction and storage tube with glass stopcock on side-arm
Z409278
Z516082
colorless tubeglass tuberound bottom
T
female joint: ST/NS14/20
Push Fit
25 mL
female ST/NS 14/20
正在寻找类似产品? 访问 产品对比指南
实验室容器与储存
实验室样品采集
玻璃耗材
批号
在文件库中查找您最近购买产品的文档。
访问文档库
1 of 9
Z173061
Solvent storage flask with glass stopcock on side-arm
Z516015
Z516104
Z516155
Z173428
Aldrich® Schlenk-type female joint cap
Z516112
Z220280
Reaction flask with glass stopcock on side-arm
Z516090
Versatile system for chemical reactions, extractions, and transfers of solids and liquids in laboratories.