主要文件
Z168068
Sigma-Aldrich
Double eyewash safety station
Includes two 32oz polyethylene bottles with covered eye cups and discharge tube
登录查看公司和协议定价
选择尺寸
1 EA
$144.00
$144.00
国内现货,预计发货时间2025年4月25日详情
选择尺寸
变更视图
1 EA
$144.00
About This Item
UNSPSC代码:
46181802
NACRES:
NB.35
$144.00
国内现货,预计发货时间2025年4月25日详情
包装
pack of 1 ea
制造商/商品名称
Bel-Art F24868-0000
一般描述
Molded plastic, wall-mountable backboard with two bottle wells, mirror, and instructions for use and maintenance in four languages (English, Spanish, French, and German). Includes two 32oz squeezable PE bottles with covered eye cups and discharge tube. Shipped empty.
历史批次信息供参考:
分析证书(COA)