Merck
Z126764
Standard point hypodermic needles, 304 stainless steel
gauge 13, L 2 in.
登录查看公司和协议定价
选择尺寸
12 EA
$296.00
$296.00
请联系客服了解存货情况
选择尺寸
变更视图
12 EA
$296.00
About This Item
UNSPSC代码:
41122002
eCl@ss:
32070500
NACRES:
NB.03
$296.00
请联系客服了解存货情况
物料
304 stainless steel needle
chrome-plated hub
包装
pack of 12 ea
制造商/商品名称
Cadence Science
长度
2 in.
计量器
13
正在寻找类似产品? 访问 产品对比指南
一般描述
Hypodermic needles are precision made and designed to ensure strict adherence with U.S. Government specification GG-N-196. Points are machine ground to achieve uniform sharpness. Precision grinding, micro-bead jet blasting, and ultrasonic cleaning assure freedom from burrs and debris. All needle cannula are fabricated of type 304 stainless steel tubing which must pass rigorous quality control prior to acceptance and use.
Standard nickle plated Luer hubs comply with Luer taper specifications ISO 594. This assures an interchangeable fit mating with Luer slips and Luer locks. Contoured hub cavity facilitates cleaning.
历史批次信息供参考:
分析证书(COA)