Merck
Z117080
Stainless steel 304 syringe needle
gauge 16, L 10 in.
登录查看公司和协议定价
选择尺寸
1 EA
$79.50
$79.50
请联系客服了解存货情况
选择尺寸
变更视图
1 EA
$79.50
About This Item
分類程式碼代碼:
41122002
eCl@ss:
32070500
NACRES:
NB.03
$79.50
请联系客服了解存货情况
描述
Standard Hub with 304 Stainless Steel Tubing
包裝
pack of 1 ea
製造商/商標名
Cadence Science
長度
10 in.
測量計
16
正在寻找类似产品? 访问 产品对比指南
一般說明
304 SS needle with chrome-plated brass Luer hub. Needle is designed with a slightly curved, deflecting tip to prevent coring during needle insertion through the septum.
Standard Hub with 304 Stainless Steel Tubing
历史批次信息供参考:
分析证书(COA)