主要文件
Z101303
Sigma-Aldrich
Aldrich® septum-inlet adapter, male
Plain, joint: ST/NS 24/40
登录查看公司和协议定价
选择尺寸
1 EA
$64.00
$64.00
请联系客服了解存货情况
选择尺寸
变更视图
1 EA
$64.00
About This Item
UNSPSC代码:
41121815
NACRES:
NB.35
$64.00
请联系客服了解存货情况
接合处
joint: ST/NS24/40
正在寻找类似产品? 访问 产品对比指南
相关类别
一般描述
Septum-inlets require septum Z565695 or Z565679 except where noted.
联系
See the Threaded Ground Glass Tutorial to see how threaded joints work.
法律信息
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
历史批次信息供参考:
分析证书(COA)
访问文档库
Questions
  1. What is the diameter of the non-glass-on-glass end?
    1 answer
    1. The outer diameter of the top tube of this non-threaded adapter is 9 mm and the bottom of the joint is approximately 24 mm outer diameter.