主要文件
SYNH200001
Synthware aluminum heat transfer block for mini vials
登录查看公司和协议定价
选择尺寸
1 EA
$100.00
$100.00
预计发货时间2025年7月11日详情
选择尺寸
变更视图
1 EA
$100.00
About This Item
UNSPSC代码:
41102404
NACRES:
NB.51
$100.00
预计发货时间2025年7月11日详情
包装
pack of 1 ea
制造商/商品名称
H200001
Kemtech H200001
相关类别
一般描述
Synthware aluminum heat transfer block is used in place of a glass sand bath in microscale experiments. It has holes to hold 3.0 mL and 5.0 mL heavy-wall reaction vials, as well as four Craig crystallization tubes, with one holder on the top for a standard thermometer.
法律信息
Synthware is a trademark of Kemtech America Inc.
历史批次信息供参考:
分析证书(COA)