主要文件
SYNB262910
Synthware bubbler with PTFE stopcock
top joint: ST/NS 29/42, hose size 10 mm
别名:
Synthware B262910
登录查看公司和协议定价
选择尺寸
1 EA
$135.00
$135.00
预计发货时间2025年6月02日详情
选择尺寸
变更视图
1 EA
$135.00
About This Item
分類程式碼代碼:
41121800
NACRES:
NB.51
$135.00
预计发货时间2025年6月02日详情
接合處
top joint: ST/NS29/42
包裝
pack of 1 ea
製造商/商標名
B262910
軟管尺寸
10 mm
相关类别
一般說明
Synthware bubbler has a small ball joint to prevent mineral oil being sucked back under sudden negative pressure in the system.
法律資訊
Synthware is a trademark of Kemtech America Inc.
历史批次信息供参考:
分析证书(COA)