主要文件
SLW1000/38D
Pyrex® Griffin beakers, low form, with printed trace code
10000 mL
别名:
Pyrex, SciLabware 1000/38D
登录查看公司和协议定价
选择尺寸
1 EA
$411.00
$411.00
预计发货时间2025年5月29日详情
选择尺寸
变更视图
1 EA
$411.00
About This Item
UNSPSC代码:
41121803
NACRES:
NB.43
$411.00
预计发货时间2025年5月29日详情
物料
borosilicate glass
white graduation
特点
low form
with printed trace code
制造商/商品名称
1000/38D
SciLabware
高度 × 直径
350 mm × 217 mm
容量
10000 mL
正在寻找类似产品? 访问 产品对比指南
一般描述
  • Complies with ISO 3819 and DIN 12331
  • Manufactured from Pyrex borosilicate glass
  • Excellent thermal performance
  • High resistance to chemical attack
  • Printed with Trace Code for downloadable batch certificate
  • Batch certificate available to download from www.scilabware.com
  • White graduations and marking spot
其他说明
Browse all glass beakers
法律信息
PYREX is a registered trademark of Corning, Inc.
历史批次信息供参考:
分析证书(COA)