主要文件
CAD4170
Stainless steel 304 syringe needle
gauge 25, L 1 in.
登录查看公司和协议定价
选择尺寸
12 EA
$629.00
$629.00
请联系客服了解存货情况
选择尺寸
变更视图
12 EA
$629.00
About This Item
UNSPSC代码:
42142600
NACRES:
NB.03
$629.00
请联系客服了解存货情况
描述
Standard Hub with 304 Stainless Steel Tubing
包装
pack of 12 ea
制造商/商品名称
Cadence Science 4170
长度
1 in.
计量器
25
正在寻找类似产品? 访问 产品对比指南
一般描述
304 SS needle with chrome-plated brass Luer hub. Needle is designed with a slightly curved, deflecting tip to prevent coring during needle insertion through the septum.
Standard Hub with 304 Stainless Steel Tubing
历史批次信息供参考:
分析证书(COA)