主要文件
BR88625
BRAND®有嘴单向旋塞
for tube I.D. 6 mm
别名:
PTFE, 旋塞
登录查看公司和协议定价
选择尺寸
1 EA
$85.20
$85.20
请联系客服了解存货情况
选择尺寸
变更视图
1 EA
$85.20
About This Item
UNSPSC代码:
41100000
NACRES:
NB.35
$85.20
请联系客服了解存货情况
物料
PTFE
包装
pack of 1 ea
制造商/商品名称
BRAND 88625
内径
3 mm
正在寻找类似产品? 访问 产品对比指南
一般描述
The BRAND® one-way stopcock with nozzles is constructed from high-quality PTFE, offering outstanding chemical resistance for various laboratory applications. This stopcock features a valve lever made of polypropylene (PP) for easy operation and control. It is equipped with integrated hose nozzles, facilitating efficient fluid transfer and connection to hoses. The robust design ensures reliable performance, making it suitable for handling aggressive chemicals and solvents.
法律信息
BRAND is a registered trademark of BRAND GMBH + CO KG
历史批次信息供参考:
分析证书(COA)