Product Name
N-Methyl-N-(trimethylsilyl)trifluoroacetamide with 1% trimethylchlorosilane derivatization grade (GC derivatization), LiChropur
Product Number
69478
Product Brand
sial
CAS Number
24589-78-4
Molecular Weight
199.25
Test
Specification
APPEARANCE (COLOR)Colorless
APPEARANCE (FORM)Liquid
PURITY (GC AREA %)≥ 97.0 %
GC MINOR COMPONENT 1 (AREA %)0.8 - 1.5 %
MINOR COMPONENT 1TRIMETHYLCHLOROSILANE
REFRACTIVE INDEX N20/D1.377 - 1.380
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE