Skip to Content
MilliporeSigma

Skip To

17876

Sodium sulfate

granulated, puriss. p.a., ACS reagent, ≥99.0%

Sign In to View Organizational & Contract Pricing.

About This Item

Linear Formula:
Na2SO4
CAS Number:
Molecular Weight:
142.04
UNSPSC Code:
12352300
PubChem Substance ID:
EC Number:
231-820-9
MDL number:


description

anhydrous

grade

ACS reagent, puriss. p.a.

assay

≥99.0% (T), ≥99.0%

quality

granulated

impurities

≤0.0005% total nitrogen (N)

loss

≤0.5% loss on ignition, 900 °C

particle size

0.2-2 mm

pH

5.2-9.2 (25 °C, 50 mg/mL in H2O)

mp

884 °C (lit.)

density

2.68 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤10 mg/kg, phosphate (PO43-): ≤10 mg/kg

cation traces

As: ≤0.1 mg/kg, Ca: ≤20 mg/kg, Cd: ≤5 mg/kg, Cr: ≤5 mg/kg, Cu: ≤5 mg/kg, Fe: ≤5 mg/kg, K: ≤50 mg/kg, Mg: ≤20 mg/kg, Mn: ≤5 mg/kg, Ni: ≤5 mg/kg, Pb: ≤5 mg/kg, Zn: ≤50 mg/kg

SMILES string

[Na+].[Na+].[O-]S([O-])(=O)=O

InChI

1S/2Na.H2O4S/c;;1-5(2,3)4/h;;(H2,1,2,3,4)/q2*+1;/p-2

InChI key

PMZURENOXWZQFD-UHFFFAOYSA-L



Still not finding the right product?

Explore all of our products under Sodium sulfate


Storage Class

13 - Non Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



The use of sodium sulfate as the globulin precipitant in the determination of proteins in blood.
Howe PE.
The Journal of Biological Chemistry, 49(1), 93-107 (1921)
How does sodium sulfate crystallize? Implications for the decay and testing of building materials.
Rodriguez-Navarro C, et al.
Cement and Concrete Research, 30(10), 1527-1534 (2000)
Teppei Adachi et al.
Inflammatory bowel diseases, 21(1), 31-39 (2014-12-02)
Expression of heat shock protein A4 (HSPA4, also called Apg-2), a member of the HSP110 family, is induced by several forms of stress. The physiological and pathological functions of HSPA4 in the intestine remain to be elucidated. We assessed HSPA4