Select a Size
| Pack Size | SKU | Availability | Price |
|---|
About This Item
Quality Segment
assay
≥98.0% (GC)
form
liquid
autoignition temp.
370 °C
potency
>5000 mg/kg LD50, oral (Rat), >5000 mg/kg LD50, skin (Rabbit)
expl. lim.
1 % (v/v)
mp
-73 °C
transition temp
flash point 46 °C
density
0.869 g/cm3 at 20 °C
storage temp.
2-30°C
SMILES string
O(CCC(C)C)C(=O)CC
InChI
1S/C8H16O2/c1-4-8(9)10-6-5-7(2)3/h7H,4-6H2,1-3H3
InChI key
XAOGXQMKWQFZEM-UHFFFAOYSA-N
Analysis Note
Density (d 20 °C/ 4 °C): 0.868 - 0.871
Identity (IR): passes test
1 of 1
This Item | |||
|---|---|---|---|
| assay ≥98.0% (GC) | assay ≥98.0% (GC) | assay ≥99.0% (GC) | assay ≥98.0% |
| density 0.869 g/cm3 at 20 °C | density 0.88 g/cm3 at 20 °C | density 0.99 g/cm3 at 20 °C | density 0.876 g/mL at 25 °C (lit.) |
| mp -73 °C | mp -73 °C | mp - | mp −78 °C (lit.) |
| expl. lim. 1 % (v/v) | expl. lim. - | expl. lim. - | expl. lim. 7.5 % |
| storage temp. 2-30°C | storage temp. 2-30°C | storage temp. 2-30°C | storage temp. - |
| potency >5000 mg/kg LD50, oral (Rat), >5000 mg/kg LD50, skin (Rabbit) | potency >5000 mg/kg LD50, oral (Rat), >5000 mg/kg LD50, skin (Rabbit) | potency 6330 mg/kg LD50, oral (Rat) | potency - |
Still not finding the right product?
Explore all of our products under Isoamyl propionate
signalword
Warning
hcodes
Hazard Classifications
Flam. Liq. 3
Storage Class
3 - Flammable liquids
wgk
WGK 2
flash_point_f
118.4 °F - closed cup
flash_point_c
48 °C - closed cup
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| 8148450100 | 04022536695518 |
| 8148450025 | 04022536695501 |




