Skip to Content
MilliporeSigma

Skip To

16-205

Anti-Phosphotyrosine Antibody, recombinant clone 4G10®, FITC conjugate

clone 4G10, Upstate®, from mouse

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
100 μg
Check Cart for Availability
$905.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
FITC conjugate
Clone:
4G10, monoclonal
Application:
ICC
Citations:
1

$905.00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Level

conjugate

FITC conjugate

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

4G10, monoclonal

species reactivity (predicted by homology)

all

manufacturer/tradename

Upstate®

technique(s)

immunocytochemistry: suitable

isotype

IgG2bκ

shipped in

wet ice

target post-translational modification

phosphorylation (pTyr)

Gene Information

human ... PID1(55022)

General description

Produced from CHO cells expressing the 4G10 antibody heavy and light chain cDNAs. Heavy chain C-terminus has a hexa-histidine tag for purification and immobilization via nickel affinity matrices. Cross-linked to fluorescein isothiocyanate (FITC). Patent pending.

Recognizes: tyrosine-phosphorylated proteins from all species

Molecular Weight: dependent upon the molecular weight of the tyrosine phosphorylated protein being detected

Species Reactivity: wide range

Immunogen

Phosphotyramine-KLH

Application

Research Category
Signaling
Research Sub Category
General Post-translation Modification
This Anti-Phosphotyrosine Antibody, recombinant clone 4G10, FITC conjugate is validated for use in IC for the detection of Phosphotyrosine.

Physical form

100µg FITC-conjugated recombinant mouse IgG2bκ in 67μl of PBS containing 0.05% sodium azide. Liquid at 4°C.

Preparation Note

9 months at 4°C

Analysis Note

routinely evaluated in immunocytochemistry Immunocytochemistry to detect phosphotyrosine-containing proteins in EGF-stimulated human A431 cells

Legal Information

4G10 is a registered trademark of Upstate Group, Inc.
UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
16-10416-10305-777
Gene Information

human ... PID1(55022)

Gene Information

human ... PID1(55022)

Gene Information

human ... PID1(55022)

Gene Information

-

conjugate

FITC conjugate

conjugate

FITC conjugate

conjugate

biotin conjugate

conjugate

-

clone

4G10, monoclonal

clone

4G10®, monoclonal

clone

4G10®, monoclonal

clone

4G10®, monoclonal

antibody form

purified immunoglobulin

antibody form

-

antibody form

purified antibody

antibody form

purified immunoglobulin

biological source

mouse

biological source

mouse

biological source

mouse

biological source

mouse

isotype

IgG2bκ

isotype

-

isotype

IgG2bκ

isotype

IgG2bκ


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Bac Viet Le et al.
Cell reports, 33(1), 108221-108221 (2020-10-08)
Synthetic lethality triggered by PARP inhibitor (PARPi) yields promising therapeutic results. Unfortunately, tumor cells acquire PARPi resistance, which is usually associated with the restoration of homologous recombination, loss of PARP1 expression, and/or loss of DNA double-strand break (DSB) end resection



Global Trade Item Number

SKUGTIN
16-20504053252583278
52214-U04061833457214

Questions

Reviews

No rating value

Active Filters