Skip to Content
MilliporeSigma

Skip To

14-844-M

PIP5K1α Protein, active, 10 µg

Active, N-terminal 6His-tagged, recombinant human PIP5K1α, full length. For use in Kinase Assays.

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
10 μg
Check Cart for Availability
$1,150.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.32
eCl@ss:
32160405
Biological source:
human
Recombinant:
expressed in baculovirus infected Sf21 cells
Mol wt:
Mw 60 kDa

$1,150.00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

human

Quality Segment

recombinant

expressed in baculovirus infected Sf21 cells

mol wt

Mw 60 kDa

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable (kinase)

UniProt accession no.

Gene Information

human ... PIP5K1A(8394)

General description

N-terminal 6His-tagged, recombinant human PIP5K1α, full length.
Product Source: Expressed by baculovirus in Sf21 insect cells

Application

Active, N-terminal 6His-tagged, recombinant human PIP5K1α, full length. For use in Kinase Assays.
Research Category
Signaling

Biochem/physiol Actions

Protein Target: PIP5K1α
Target Sub-Family: PIPK

Packaging

Also available in 250μg size as catalog # 14-844M - Call for pricing and availability.

Physical form

10µg of enzyme in 50mM Tris/HCl pH8.0, 300mM NaCl, 0.1mM EGTA, 0.03% Brij-35, 270mM sucrose, 0.2mM PMSF, 1mM benzamidine, 0.1% 2-mercaptoethanol. Frozen solution.
Immobilized metal affinity chromatography (IMAC)

Preparation Note

6 months at -70°C

Analysis Note

Routinely evaluated in the PIProfiler.

Other Notes

For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
14-777-M14-552-M14-565-M
technique(s)

activity assay: suitable (kinase)

technique(s)

activity assay: suitable (kinase)

technique(s)

activity assay: suitable (kinase)

technique(s)

activity assay: suitable (kinase)

biological source

human

biological source

human

biological source

human

biological source

human

recombinant

expressed in baculovirus infected Sf21 cells

recombinant

expressed in baculovirus infected Sf21 cells

recombinant

expressed in baculovirus infected Sf21 cells

recombinant

-

mol wt

Mw 60 kDa

mol wt

Mw 69.3 kDa

mol wt

Mw 78.4 kDa

mol wt

Mw 37.9 kDa

Gene Information

human ... PIP5K1A(8394)

Gene Information

-

Gene Information

human ... BTK(695)

Gene Information

-

UniProt accession no.

Q99755, U78577

UniProt accession no.

P53350, Q07832 (mouse), Q62673 (rat)

UniProt accession no.

Q06187

UniProt accession no.

Q8TDX7


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Skin Sens. 1

Storage Class

12 - Non Combustible Liquids

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
14-84404053252277160
14-844M04053252669231

Questions

Reviews

No rating value

Active Filters