Merck
Suppression of experimental autoimmune encephalomyelitis by ultraviolet light is not mediated by isomerization of urocanic acid.
BMC neuroscience (2017-01-07)
Amy A Irving, Steven J Marling, Lori A Plum, Hector F DeLuca
ABSTRAKT
Ultraviolet B irradiation confers strong resistance against experimental autoimmune encephalomyelitis, a model of multiple sclerosis. This protection by ultraviolet B is independent of vitamin D production but causes isomerization of urocanic acid, a naturally occurring immunosuppressant. To determine whether UCA isomerization from trans to cis is responsible for the protection against experimental autoimmune encephalomyelitis afforded by ultraviolet B, trans- or cis-urocanic acid was administered to animals and their disease progression was monitored. Disease incidence was reduced by 74% in animals exposed to ultraviolet B, and skin cis-urocanic acid levels increased greater than 30%. However, increasing skin cis-urocanic acid levels independent of ultraviolet B was unable to alter disease onset or progression. It is unlikely that urocanic acid isomerization is responsible for the ultraviolet B-mediated suppression of experimental autoimmune encephalomyelitis. Additional work is needed to investigate alternative mechanisms by which UVB suppresses disease.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
cis-Urocanic acid, ≥98% (HPLC), solid (fluffy)