Merck
Vibrational spectroscopy of resveratrol.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2007-04-10)
Ferenc Billes, Ildikó Mohammed-Ziegler, Hans Mikosch, Erno Tyihák
ABSTRAKT
In this article the authors deal with the experimental and theoretical interpretation of the vibrational spectra of trans-resveratrol (3,5,4'-trihydroxy-trans-stilbene) of diverse beneficial biological activity. Infrared and Raman spectra of the compound were recorded; density functional calculations were carried out resulting in the optimized geometry and several properties of the molecule. Based on the calculated force constants, a normal coordinate analysis yielded the character of the vibrational modes and the assignment of the measured spectral bands.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Pinosylvin, ≥97.0% (HPLC)