- Novel thrombin inhibitors incorporating non-basic partially saturated heterobicyclic P1-arginine mimetics.
Novel thrombin inhibitors incorporating non-basic partially saturated heterobicyclic P1-arginine mimetics.
Bioorganic & medicinal chemistry letters (2003-03-06)
Lucija Peterlin-Masic, Gregor Mlinsek, Tomaz Solmajer, Alenka Trampus-Bakija, Mojca Stegnar, Danijel Kikelj
PMID12617892
ABSTRAKT
The design, synthesis and biological activity of non-covalent thrombin inhibitors incorporating 4,5,6,7-tetrahydroindazole, 2-methyl-4,5,6,7-tetrahydroindazole, 4,5,6,7-tetrahydroisoindole, 5,6,7,8-tetrahydroquinazoline and 5,6,7,8-tetrahydroquinazolin-2-amine as novel, partially saturated, heterobicyclic P(1)-arginine side-chain mimetics is described. The binding mode of the most potent candidate in the series co-crystallized with human alpha-thrombin, which exhibited an in vitro K(i) of 140nM and more that 478-fold selectivity against trypsin, is discussed.
MATERIAŁY