Merck
  • The complete genome of Bacillus amyloliquefaciens subsp. plantarum CAU B946 contains a gene cluster for nonribosomal synthesis of iturin A.
The complete genome of Bacillus amyloliquefaciens subsp. plantarum CAU B946 contains a gene cluster for nonribosomal synthesis of iturin A.
Journal of bacteriology (2012-03-13)
Jochen Blom, Christian Rueckert, Ben Niu, Qi Wang, Rainer Borriss
ABSTRAKT
The genome of the rhizobacterium Bacillus amyloliquefaciens subsp. plantarum CAU B946 was 4.02 Mb in size and harbored 3,823 genes (coding sequences [CDS]). Nine giant gene clusters were dedicated to nonribosomal synthesis of antimicrobial compounds. Remarkably, strain CAU B946 possessed a gene cluster involved in synthesis of iturin A.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Iturin A from Bacillus subtilis, ≥95% (HPLC)