Merck
  • Pharmacodynamic and antineoplastic activity of BI 836845, a fully human IGF ligand-neutralizing antibody, and mechanistic rationale for combination with rapamycin.
Pharmacodynamic and antineoplastic activity of BI 836845, a fully human IGF ligand-neutralizing antibody, and mechanistic rationale for combination with rapamycin.
Molecular cancer therapeutics (2013-12-04)
Katrin Friedbichler, Marco H Hofmann, Monika Kroez, Elinborg Ostermann, Herbert R Lamche, Christian Koessl, Eric Borges, Michael N Pollak, Günther Adolf, Paul J Adam
ABSTRAKT
Insulin-like growth factor (IGF) signaling is thought to play a role in the development and progression of multiple cancer types. To date, therapeutic strategies aimed at disrupting IGF signaling have largely focused on antibodies that target the IGF-I receptor (IGF-IR). Here, we describe the pharmacologic profile of BI 836845, a fully human monoclonal antibody that utilizes an alternative approach to IGF signaling inhibition by selectively neutralizing the bioactivity of IGF ligands. Biochemical analyses of BI 836845 demonstrated high affinity to human IGF-I and IGF-II, resulting in effective inhibition of IGF-induced activation of both IGF-IR and IR-A in vitro. Cross-reactivity to rodent IGFs has enabled rigorous assessment of the pharmacologic activity of BI 836845 in preclinical models. Pharmacodynamic studies in rats showed potent reduction of serum IGF bioactivity in the absence of metabolic adverse effects, leading to growth inhibition as evidenced by reduced body weight gain and tail length. Moreover, BI 836845 reduced the proliferation of human cell lines derived from different cancer types and enhanced the antitumor efficacy of rapamycin by blocking a rapamycin-induced increase in upstream signaling in vitro as well as in human tumor xenograft models in nude mice. Our data suggest that BI 836845 represents a potentially more effective and tolerable approach to the inhibition of IGF signaling compared with agents that target the IGF-I receptor directly, with potential for rational combinations with other targeted agents in clinical studies.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Anti-IGF-IR (Ab-1) Mouse mAb (αIR3), lyophilized, clone αIR3, Calbiochem®