Merck
Valco® Precision Sampling Syringe, Series C, Removable needle volume 1000 μL, bevel tip stainless steel needle (point style 2), needle L × O.D. × I.D. 2 in. × 0.029 in. × 0.012 in.
Valco® Precision Sampling Syringe, Series C, Removable needle
Valco® Precision Sampling Syringe, Series C, Removable needle volume 1000 μL, bevel tip stainless steel needle (point style 2), needle L × O.D. × I.D. 2 in. × 0.029 in. × 0.012 in.
Products
Kategoria produktu
Producent
Długość (cale)
Objętość (µL)
Kształt igły