Wybierz wielkość
| Do Państwa/SKU | Dostępność | Cena netto |
|---|---|---|
1 ea | Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności | 9850,00 zł |
Informacje o tej pozycji
9850,00 zł
agency
suitable for DIN 38407-42, suitable for EPA 1633, suitable for EPA 533, suitable for EPA 537.1, suitable for EPA OTM-45, suitable for GB 31604.35-2016, suitable for ISO 21675 2019, suitable for ISO 25101, suitable for ISO/CEN 15968-2010
reg. compliance
suitable for FDA C-010.02
technique(s)
solid phase extraction (SPE): suitable
application(s)
PFAS testing
food and beverages
compatibility
for use with Visiprep 24-port model
General description
Note: The Visidry drying attachment cannot be used to dry 12mL, 20mL, or 60mL SPE tubes.
Legal Information
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description for use with Visiprep 24-port model | description for use with Visiprep 12-port model | description for use with 24-port model, includes base, 2 support rods, center plate, plate for 10 mm test tubes, 8 retaining clips | description for use with 24-port model (standard and DL), pk of 2 |
| application(s) PFAS testing | application(s) PFAS testing | application(s) food and beverages | application(s) food and beverages |
| technique(s) solid phase extraction (SPE): suitable | technique(s) solid phase extraction (SPE): suitable | technique(s) solid phase extraction (SPE): suitable | technique(s) solid phase extraction (SPE): suitable |
| compatibility for use with Visiprep 24-port model | compatibility for use with Visiprep 12-port model | compatibility for use with 24-port model | compatibility for use with 24-port model (standard and DL) |
| agency suitable for DIN 38407-42, suitable for EPA 533, suitable for EPA OTM-45, suitable for ISO 21675 2019, suitable for ISO/CEN 15968-2010, suitable for EPA 1633, suitable for GB 31604.35-2016, suitable for EPA 537.1 | agency suitable for DIN 38407-42, suitable for EPA 1633, suitable for EPA 533, suitable for EPA 537.1, suitable for EPA OTM-45, suitable for GB 31604.35-2016, suitable for ISO 21675 2019, suitable for ISO 25101, suitable for ISO/CEN 15968-2010 | agency - | agency - |
| reg. compliance suitable for FDA C-010.02 | reg. compliance suitable for FDA C-010.02 | reg. compliance - | reg. compliance - |
ASTM8421 | CEN-TS15968-2010, DIN17681-1, DIN17681-2, DIN17892, DIN38407-42, DIN38414-14, EPA1633, EPA537_1, EPAPlasticContainerMethod, EPAmethod533, FDAC-010_02, FDAC-010_03, GB31604_35-2016, HJ1333-2023, HJ1334-2023, ISO21675_2019, ISO25101, MoEE3457 | - | - |
Still not finding the right product?
Explore all of our products under Visidry Drying Attachment
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?