Kluczowe dokumenty
56741-U
Supelco
Stainless Steel 1/16 in. Capillary Tubing
L × O.D. × I.D. 0.5 m × 1/16 in. × 0.040 in.
About This Item
Kod UNSPSC:
41115700
eCl@ss:
32119290
NACRES:
SB.52
Materiały
none band
dł. × śr. zewn. × śr. wewn.
0.5 m × 1/16 in. × 0.040 in.
Opis ogólny
316 stainless steel, 1/16 in. O.D. 1 m length is coiled, all other dimensions are straight.
Polecane produkty
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)