Zmień widok
| Do Państwa/SKU | Dostępność | Cena netto |
|---|---|---|
12 ea | Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności | 845,00 zł |
Informacje o tej pozycji
UNSPSC Code:
41122001
NACRES:
NB.43
845,00 zł
Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności
Pomoc techniczna
Pozwól nam pomócmaterial
white PTFE valve
packaging
pkg of 12 ea
application(s)
general analytical
compatibility
for use with 18/400 mm thread
General description
These valves are very easy to use.
1. Push the green button to open the valve
2. Insert the needle through the septum and take sample
3. Remove the needle and push to close
Benefits include:
Silicone septa, red
1. Push the green button to open the valve
2. Insert the needle through the septum and take sample
3. Remove the needle and push to close
Benefits include:
- PTFE construction for inertness. PTFE is useful when working when acids and organic solvents.
- Good sealing ability for containing most volalite liquids and gases at low pressures
- Mininert valves can be used at temperatures up to 105 °F.
- 0.036 in. (0.9 mm) opening accepts up to 20-gauge needle
Silicone septa, red
Application
Mininert valves are leak-tight closures for vials and other containers. They are available in both crimp and screw thread versions. Only PTFE and glass are in contact with the contents of the container. They are the ideal closure for the following applications:
- calibration standards
- air- or moisture-sensitive fluids
- derivatizating reagents
- volatile chemicals
Other Notes
The maximum temperature that Mininert Valves can be used at is 105° F. When Mininert Valves are used at high temperatures, returning them to room temperature may cause the valve to leak slightly.
Legal Information
Mininert is a registered trademark of Valco Instruments Co. Inc.
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description for use with 18/400 mm thread, pkg of 12 ea | description for use with 13/425 mm thread, pkg of 12 ea | description for use with 15/425 mm thread, pkg of 12 ea | description for use with 24/400 mm thread, pkg of 12 ea |
| material white PTFE valve | material white PTFE valve | material white PTFE valve | material white PTFE valve |
| compatibility for use with 18/400 mm thread | compatibility for use with 13/425 mm thread | compatibility for use with 15/425 mm thread | compatibility for use with 24/400 mm thread |
| packaging pkg of 12 ea | packaging pkg of 12 ea | packaging pkg of 12 ea | packaging pkg of 12 ea |
Still not finding the right product?
Explore all of our products under Mininert® Valve, screw thread
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?