Wybierz wielkość
| Rozmiar/SKU | Dostępność | Cena netto |
|---|---|---|
12 ea | Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności | 820,00 zł |
Informacje o tej pozycji
820,00 zł
material
white PTFE valve
packaging
pkg of 12 ea
application(s)
general analytical
compatibility
for use with 13/425 mm thread
General description
1. Push the green button to open the valve
2. Insert the needle through the septum and take sample
3. Remove the needle and push to close
Benefits include:
- PTFE construction for inertness. PTFE is useful when working when acids and organic solvents.
- Good sealing ability for containing most volalite liquids and gases at low pressures
- Mininert valves can be used at temperatures up to 105 °F.
- 0.036 in. (0.9 mm) opening accepts up to 20-gauge needle
Silicone septa, red
Application
- calibration standards
- air- or moisture-sensitive fluids
- derivatizating reagents
- volatile chemicals
Other Notes
Legal Information
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description for use with 13/425 mm thread, pkg of 12 ea | description for use with 15/425 mm thread, pkg of 12 ea | description for use with 18/400 mm thread, pkg of 12 ea | description for use with 24/400 mm thread, pkg of 12 ea |
| material white PTFE valve | material white PTFE valve | material white PTFE valve | material white PTFE valve |
| compatibility for use with 13/425 mm thread | compatibility for use with 15/425 mm thread | compatibility for use with 18/400 mm thread | compatibility for use with 24/400 mm thread |
| packaging pkg of 12 ea | packaging pkg of 12 ea | packaging pkg of 12 ea | packaging pkg of 12 ea |
| application(s) general analytical | application(s) general analytical | application(s) general analytical | application(s) environmental |
Still not finding the right product?
Explore all of our products under Mininert® Valve, screw thread
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?