Wybierz wielkość
| Rozmiar/SKU | Dostępność | Cena netto |
|---|---|---|
1 ea | Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności | 531,00 zł |
Informacje o tej pozycji
531,00 zł
material
bevel tip stainless steel needle (point style 2)
description
Series A-2
feature
removable needle Series A2
packaging
pkg of 1 ea
manufacturer/tradename
VICI 050023
needle L × O.D. × I.D.
2 in. × 0.028 in. × 0.005 in.
volume
25 μL
application(s)
general analytical
General description
- Push-button valve - allows instantaneous injection
- Smaller volumes - great for small volatile samples
Legal Information
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description volume 25 μL, bevel tip stainless steel needle (point style 2), needle L × O.D. × I.D. 2 in. × 0.028 in. × 0.005 in. | description volume 250 μL, bevel tip stainless steel needle (point style 2), needle L × O.D. × I.D. 2 in. × 0.029 in. × 0.012 in. | description volume 5 mL, bevel tip stainless steel needle (point style 2), needle L × O.D. × I.D. 2 in. × 0.029 in. × 0.012 in. | description volume 500 μL, bevel tip stainless steel needle (point style 2), needle L × O.D. × I.D. 2 in. × 0.029 in. × 0.012 in. |
| material bevel tip stainless steel needle (point style 2) | material bevel tip stainless steel needle (point style 2) | material bevel tip stainless steel needle (point style 2) | material bevel tip stainless steel needle (point style 2) |
| packaging pkg of 1 ea | packaging pkg of 1 ea | packaging pkg of 1 ea | packaging pkg of 1 ea |
| manufacturer/tradename VICI 050023 | manufacturer/tradename VICI 050031 | manufacturer/tradename VICI 050035 | manufacturer/tradename VICI 050032 |
| volume 25 μL | volume 250 μL | volume 5 mL | volume 500 μL |
| application(s) general analytical | application(s) general analytical | application(s) general analytical | application(s) general analytical |
| needle L × O.D. × I.D. 2 in. × 0.028 in. × 0.005 in. | needle L × O.D. × I.D. 2 in. × 0.029 in. × 0.012 in. | needle L × O.D. × I.D. 2 in. × 0.029 in. × 0.012 in. | needle L × O.D. × I.D. 2 in. × 0.029 in. × 0.012 in. |
Still not finding the right product?
Explore all of our products under Valco® Precision Sampling Syringe, Series A-2, Removable needle
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?