Wybierz wielkość
| Rozmiar/SKU | Dostępność | Cena netto |
|---|---|---|
10 ea | Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności | 170,00 zł |
Informacje o tej pozycji
170,00 zł
material
PTFE back ferrule
packaging
pkg of 10 ea
technique(s)
gas chromatography (GC): suitable using :Suitable
I.D.
1/8 in.
fitting
SWAGELOK for 1/8 in. tubing
compatibility
for use with Purge Traps (back ferrule)
General description
Application
Legal Information
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description PTFE back ferrule, SWAGELOK for 1/8 in. tubing | description PTFE front ferrule, SWAGELOK for 1/8 in. tubing | description PTFE back ferrule, SWAGELOK for 1/16 in. tubing | description PTFE front ferrule, SWAGELOK for 1/4 in. tubing |
| I.D. 1/8 in. | I.D. 1/8 in. | I.D. - | I.D. 1/4 in. |
| material PTFE back ferrule | material PTFE front ferrule | material PTFE back ferrule | material PTFE front ferrule |
| technique(s) gas chromatography (GC): suitable using :Suitable | technique(s) gas chromatography (GC): suitable using :Suitable | technique(s) gas chromatography (GC): suitable using :Suitable | technique(s) gas chromatography (GC): suitable using :Suitable |
| fitting SWAGELOK for 1/8 in. tubing | fitting SWAGELOK for 1/8 in. tubing | fitting SWAGELOK for 1/16 in. tubing | fitting SWAGELOK for 1/4 in. tubing |
| packaging pkg of 10 ea | packaging pkg of 10 ea | packaging pkg of 5 ea | packaging pkg of 10 ea |
| compatibility for use with Purge Traps (back ferrule) | compatibility for use with Purge Traps (front ferrule) | compatibility - | compatibility - |
Still not finding the right product?
Explore all of our products under Swagelok® PTFE Ferrules
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?