Przejdź do
22058
SWAGELOK® PTFE Ferrules
PTFE front ferrule, SWAGELOK for 1/8 in. tubing
Wybierz wielkość
Zmień widok
Rozmiar/SKUDostępnośćCena netto
10 ea
Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności
244,00 zł
Informacje o tej pozycji
UNSPSC Code:
41115717
NACRES:
SB.54
244,00 zł
Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności
​
Złóż zamówienie na produkt na zamówienie
Pomoc techniczna
Pozwól nam pomóc
material
PTFE front ferrule
packaging
pkg of 10 ea
technique(s)
gas chromatography (GC): suitable using :Suitable
I.D.
1/8 in.
fitting
SWAGELOK for 1/8 in. tubing
compatibility
for use with Purge Traps (front ferrule)
General description
Use PTFE ferrules instead of brass or stainless steel ferrules when attaching fittings to glass tubing, or to flexible tubing (such as PTFE or Tygon).
Swage type fittings are one of the most popular connectors which are available in metal and plastic forms; they are very useful for connecting similar or dissimilar materials, like metal to glass or metal to plastic. The general swage tubing coupler has four parts namely the body, into which the tube is placed, two ferrules that is front ferrule and back ferrule and lastly a packing nut.
Application
Swagelok® PTFE ferrules maybe used, for making all the connections between glass and PTFE present in the containment vessels, piping and sampling equipment in a bench scale biofilter system, in an experimental study investigating the effects of periods of non-use on biofilter performance.
Legal Information
SWAGELOK is a registered trademark of Swagelok Company
Porównaj podobne pozycje
Wyświetl pełne porównanie
Pokaż różnice
1 of 1
Ta pozycja
220592206922027
description
PTFE front ferrule, SWAGELOK for 1/8 in. tubing
description
PTFE back ferrule, SWAGELOK for 1/8 in. tubing
description
PTFE back ferrule, SWAGELOK for 1/16 in. tubing
description
SWAGELOK® 403-1, stainless steel, 1/4 in. SWAGELOK
I.D.
1/8 in.
I.D.
1/8 in.
I.D.
-
I.D.
-
material
PTFE front ferrule
material
PTFE back ferrule
material
PTFE back ferrule
material
stainless steel
fitting
SWAGELOK for 1/8 in. tubing
fitting
SWAGELOK for 1/8 in. tubing
fitting
SWAGELOK for 1/16 in. tubing
fitting
1/4 in. SWAGELOK
technique(s)
gas chromatography (GC): suitable using :Suitable
technique(s)
gas chromatography (GC): suitable using :Suitable
technique(s)
gas chromatography (GC): suitable using :Suitable
technique(s)
gas chromatography (GC): suitable using :Suitable
compatibility
for use with Purge Traps (front ferrule)
compatibility
for use with Purge Traps (back ferrule)
compatibility
-
compatibility
-
packaging
pkg of 10 ea
packaging
pkg of 10 ea
packaging
pkg of 5 ea
packaging
pkg of 5 ea
Still not finding the right product?
Explore all of our products under Swagelok® PTFE Ferrules
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)