Przejdź do
Z768642
Nalgene® square graduated media bottle
with cap, PET, sterile, capacity 250 mL
Synonim(y):
media storage bottle, square plastic bottle, Nalgene 342040-0250, media container, reagent bottle
Wybierz wielkość
Zmień widok
Rozmiar/SKUDostępnośćCena netto
Informacje o tej pozycji
NACRES:
NB.17
UNSPSC Code:
41121800
Capacity:
250 mL
Material:
PET , square bottle
Sterility:
sterile
Feature:
graduations, with cap
Packaging:
pack of 60 ea (2 x, pack of 30)
Informacje o cenach i dostępności nie są obecnie dostępne.
Pomoc techniczna
Pozwól nam pomóc
material
PET , square bottle
joint
Joints threaded neck (38-430)
sterility
sterile
feature
graduations, with cap
packaging
pack of 60 ea (2 x, pack of 30)
manufacturer/tradename
Nalgene® 342040-0250
capacity
250 mL
General description
Nalgene™ Square PET media bottles offer sterility, lightweight construction, and shatter resistance. They are specifically designed for use in filling lines.
Features and Benefits
  • Has molded-in graduations and neck-ring.
  • Non-cytotoxic, non-hemolytic, and non-pyrogenic.
  • Exhibits exceptional gas barrier properties.
  • Gamma irradiated to 10^-6 to achieve sterility assurance level (SAL).
  • Composed of material free from ADC (animal derived component).
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Porównaj podobne pozycje
Wyświetl pełne porównanie
Pokaż różnice
1 of 1
Ta pozycja
Z768650Z768669Z768634
description
with cap, PET, sterile, capacity 250 mL
description
with cap, PET, sterile, capacity 650 mL
description
with cap, PET, sterile, capacity 1000 mL
description
with cap, PET, sterile, capacity 125 mL
capacity
250 mL
capacity
650 mL
capacity
1000 mL
capacity
125 mL
feature
graduations, with cap
feature
graduations, with cap
feature
graduations, with cap
feature
with cap, graduations
sterility
sterile
sterility
sterile
sterility
sterile
sterility
sterile
material
square bottle, PET
material
square bottle, PET
material
PET , square bottle
material
PET , square bottle
packaging
pack of 60 ea (2 x, pack of 30)
packaging
pack of 48 ea (2 x pack of 24)
packaging
pack of 24 ea (2 x pack of 12)
packaging
pack of 96 ea (4 x pack of 24)
manufacturer/tradename
Nalgene® 342040-0250
manufacturer/tradename
Nalgene® 842040-0650
manufacturer/tradename
Nalgene 842040-1000
manufacturer/tradename
Nalgene 342040-0125
Still not finding the right product?
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)