Wybierz wielkość
| Rozmiar/SKU | Dostępność | Cena netto |
|---|---|---|
48 ea | Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności | 1440,00 zł |
Informacje o tej pozycji
1440,00 zł
material
PET , square bottle
joint
Joints threaded neck (38-430)
sterility
sterile
feature
graduations, with cap
packaging
pack of 48 ea (2 x pack of 24)
manufacturer/tradename
Nalgene® 842040-0650
capacity
650 mL
General description
Features and Benefits
- Has molded-in graduations and neck-ring.
- Non-cytotoxic, non-hemolytic, and non-pyrogenic.
- Exhibits exceptional gas barrier properties.
- Gamma irradiated to 10^-6 to achieve sterility assurance level (SAL).
- Composed of material free from ADC (animal derived component).
Legal Information
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description with cap, PET, sterile, capacity 650 mL | description without cap, PET, sterile, capacity 125 mL | description with cap, PET, sterile, capacity 1000 mL | description capacity 650 mL |
| sterility sterile | sterility sterile | sterility sterile | sterility sterile; γ-irradiated |
| feature with cap, graduations | feature graduations, without cap | feature graduations, with cap | feature graduations: no, with cap |
| capacity 650 mL | capacity 125 mL | capacity 1000 mL | capacity 650 mL |
| material square bottle, PET | material square bottle, PET | material square bottle, PET | material high-density polyethylene cap, square bottle, transparent PETG |
| packaging pack of 48 ea (2 x pack of 24) | packaging pack of 96 ea (4 x pack of 24) | packaging pack of 24 ea (2 x pack of 12) | packaging pack of 48 ea (2 x pack of 24) |
| manufacturer/tradename Nalgene® 842040-0650 | manufacturer/tradename Nalgene 842044-0125 | manufacturer/tradename Nalgene 842040-1000 | manufacturer/tradename Nalgene® 342020-9500 |
Still not finding the right product?
Explore all of our products under Nalgene® square graduated media bottle
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?