Zmień widok
| Do Państwa/SKU | Dostępność | Cena netto |
|---|---|---|
500 ea | Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności | 211,00 zł |
Informacje o tej pozycji
NACRES:
NB.35
UNSPSC Code:
41122410
Manufacturer/tradename:
Diversified Biotech FOIL-4000
Sterility:
non-sterile
Packaging:
pack of 500 sheets
211,00 zł
Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności
Pomoc techniczna
Pozwól nam pomócsterility
non-sterile
packaging
pack of 500 sheets
manufacturer/tradename
Diversified Biotech FOIL-4000
L × W
5 in. × 5 in.
General description
Speedy-Foil is pre-cut aluminum squares interleved with tissue paper for conveniently and quickly covering lab vessels.
Features and Benefits
- Faster, easier and aseptic: grab and cover
- Pre-cut to fit common laboratory containers
- No scissors and less waste
- 1 mil thick for a sharp, crisp dead-fold providing a tighter fit
- Each square is interleaved for physical separation
- Meets FDA requirements for direct contact with pharmaceuticals
- RNase and DNase free
Legal Information
Speedy-Foil is a registered trademark of Diversified Biotech
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description L × W 5 in. × 5 in., pack of 500 sheets | description L × W 3 in. × 3 in., pack of 500 sheets | description L × W 4 in. × 4 in., pack of 500 sheets | description L × W 2 in. × 2 in., pack of 500 sheets |
| manufacturer/tradename Diversified Biotech FOIL-4000 | manufacturer/tradename Diversified Biotech FOIL-2000 | manufacturer/tradename Diversified Biotech FOIL-3000 | manufacturer/tradename Diversified Biotech FOIL-1000 |
| sterility non-sterile | sterility non-sterile | sterility non-sterile | sterility - |
| packaging pack of 500 sheets | packaging pack of 500 sheets | packaging pack of 500 sheets | packaging pack of 500 sheets |
| L × W 5 in. × 5 in. | L × W 3 in. × 3 in. | L × W 4 in. × 4 in. | L × W 2 in. × 2 in. |
Still not finding the right product?
Explore all of our products under Speedy-Foil® aluminium squares interleaved with paper
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?