Z741695
Sartorius mLINE® mechanical pipettor Multipacks
Volume: Multiple (20 μl, 200 μl, 1,000 μl)
About This Item
Kod UNSPSC:
41121500
opis
Includes stand and 2 x 200 μl and 1000 μl Tip trays
Właściwości
manual
single-channel
opakowanie
pack of 3 pieces
producent / nazwa handlowa
Sartorius LH-725662
pojemność
1,000 μL
20 μL
200 μL
Opis ogólny
The mLINE® mechanical pipette Multipacks offer an affordable option to get a set of quality pipettes with useful accessories in a convenient package of your choice. Whether you are renewing your existing pipettes or setting up a new pipetting station, these packs will fulfill your need. Sartorius′ most advanced mechanical pipette family - the mLINE[ - offers excellent ergonomics, performance and safety in manual pipetting. It is particularly designed for repetitive and long-lasting pipetting in order to prevent work related hand, arm and shoulder disorders or Repetitive Strain Injury (RSI). mLINE is recommended by health and safety officers around the globe.
Informacje prawne
mLINE is a registered trademark of Sartorius Biohit Liquid Handling Oy
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)