Kluczowe dokumenty
Z704075
Metal Polymer Nanocomposites
About This Item
ISBN-10:
0-47147131-3
ISBN-13:
978-0-47147131-8
Kod UNSPSC:
55101509
informacje o publikacji
L. Nicolais and G. Carotenuto, John Wiley & Sons, 2004, 320 pp., hard cover
Opis ogólny
A unique guide to an essential area of nanoscience. Interest in nano-sized metals has increased greatly due to their special characteristics and suitability for a number of advanced applications. As technology becomes more refined-including the ability to effectively manipulate and stabilize metals at the nanoscale-these materials present ever-more workable solutions to a growing range of problems. The coverage includes: Chemical and physical properties of nano-sized metals; Different approaches to the synthesis of metal-polymer nanocomposites (MPN); Advanced characterization techniques and methods for study of MPN; Real-world applications, including color filters, polarizers, optical sensors, nonlinear optical devices, and more.
Spis treści
1. Physical and chemical properties of nanosized metal particles
2. Metal containing polymers: cryochemical synthesis, structure and physico-chemical properties .
3. Controlled pyrolysis of metal-containing precursors as a way for synthesis of metallopolymer nanocomposites
4. Nanostructured polymeric nanoreactors for metal nanoparticle formation .
5. Metal-polymer nanocomposite synthesis: Novel Ex Situ and In Situ approaches
6. Plamon absorption of embedded nanoparticles .
7. Magnetooptic of granular materials and new optical methods of magnetic nanoparticles and nanostructures imaging .
8. Optical extinction of metal nanoparticles synthesized in polymer by ion implantation .
9. Optically anisotropic metal-polymer nanocomposites
Index.
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)