Przejdź do
Z364576
Nalgene® media bottles, style 2020
capacity 500 mL
Synonim(y):
bottle nalgene, nalgene containers, sterile bottles, Nalgene 342020-0500, media container, reagent bottle
Wybierz wielkość
Zmień widok
Rozmiar/SKUDostępnośćCena netto
40 ea
Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności
1860,00 zł
Informacje o tej pozycji
NACRES:
NB.17
UNSPSC Code:
41121800
Capacity:
500 mL
Material:
high-density polyethylene cap, square bottle, transparent PETG
Sterility:
sterile; γ-irradiated
Feature:
graduations: no, with cap
Packaging:
pack of 40 ea (2 x pack of 20)
1860,00 zł
Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności
Pomoc techniczna
Pozwól nam pomóc
material
high-density polyethylene cap, square bottle, transparent PETG
joint
Joints threaded neck (38-430mm)
description
Not autoclavable
sterility
sterile; γ-irradiated
feature
graduations: no, with cap
packaging
pack of 40 ea (2 x pack of 20)
manufacturer/tradename
Nalgene® 342020-0500
capacity
500 mL
General description
Media bottles (square). Same materials and design as Style 2019, but without shrink band around neck.
Features and Benefits
  • Extensive validation documentation is available
  • Produced from durable, break-resistant polyethylene terephthalate copolyester, glycol modified (PETG) that enhance reduced permeability to CO2/O2.
  • Tamper evident shrink sleeves used along with moulded in graduations and neck ring.
  • Non-pyrogenic and non-cytotoxic
  • Animal derived component free (ADCF) material for bottle and closure material.
  • Leakproof and transparent
  • Devoid of costly washing, depyrogenation steps
  • Prolonged storage of pH-sensitive liquids such as culture media
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Porównaj podobne pozycje
Wyświetl pełne porównanie
Pokaż różnice
1 of 1
Ta pozycja
Z364533Z364568Z364592
description
capacity 500 mL
description
capacity 60 mL
description
capacity 250 mL
description
capacity 1000 mL
sterility
sterile; γ-irradiated
sterility
sterile; γ-irradiated
sterility
sterile; γ-irradiated
sterility
sterile; γ-irradiated
feature
graduations: no, with cap
feature
with cap, graduations: no
feature
graduations: no, with cap
feature
with cap, graduations: no
capacity
500 mL
capacity
60 mL
capacity
250 mL
capacity
1000 mL
material
square bottle, high-density polyethylene cap, transparent PETG
material
high-density polyethylene cap, transparent PETG , square bottle
material
transparent PETG , high-density polyethylene cap, square bottle
material
transparent PETG , high-density polyethylene cap, square bottle
packaging
pack of 40 ea (2 x pack of 20)
packaging
pack of 200 ea (5 xpack of 40)
packaging
pack of 60 ea (2 xpack of 30)
packaging
pack of 24 ea (2 xpack of 12)
joint
Joints threaded neck (38-430mm)
joint
Joints threaded neck (24-415mm)
joint
Joints threaded neck (38-430mm)
joint
Joints threaded neck (38-430mm)
Still not finding the right product?
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)