Wybierz wielkość
| Rozmiar/SKU | Dostępność | Cena netto |
|---|---|---|
200 ea | Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności | 3220,00 zł 2737,00 zł |
Informacje o tej pozycji
2737,00 zł
Cena katalogowa3220,00 złZaoszczędź 15%material
high-density polyethylene cap, square bottle, transparent PETG
joint
Joints threaded neck (24-415mm)
description
Not autoclavable
sterility
sterile; γ-irradiated
feature
graduations: no, with cap
packaging
pack of 200 ea (5 xpack of 40)
manufacturer/tradename
Nalgene® 342020-0060
capacity
60 mL
General description
Features and Benefits
- Extensive validation documentation is available
- Produced from durable, break-resistant polyethylene terephthalate copolyester, glycol modified (PETG) that enhance reduced permeability to CO2/O2.
- Tamper evident shrink sleeves used along with moulded in graduations and neck ring.
- Non-pyrogenic and non-cytotoxic
- Animal derived component free (ADCF) material for bottle and closure material.
- Leakproof and transparent
- Devoid of costly washing, depyrogenation steps
- Prolonged storage of pH-sensitive liquids such as culture media
Legal Information
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description capacity 60 mL | description capacity 2000 mL | description capacity 1000 mL | description capacity 125 mL |
| sterility sterile; γ-irradiated | sterility sterile; γ-irradiated | sterility sterile; γ-irradiated | sterility sterile; γ-irradiated |
| feature graduations: no, with cap | feature graduations: no, with cap | feature graduations: no, with cap | feature graduations: no, with cap |
| capacity 60 mL | capacity 2000 mL | capacity 1000 mL | capacity 125 mL |
| material square bottle, transparent PETG , high-density polyethylene cap | material square bottle, transparent PETG , high-density polyethylene cap | material high-density polyethylene cap, square bottle, transparent PETG | material high-density polyethylene cap, square bottle, transparent PETG |
| packaging pack of 200 ea (5 xpack of 40) | packaging pack of 12 ea (2 xpack of 6) | packaging pack of 24 ea (2 xpack of 12) | packaging pack of 96 ea (4 xpack of 24) |
| joint Joints threaded neck (24-415mm) | joint Joints threaded neck (53B) | joint Joints threaded neck (38-430mm) | joint Joints threaded neck (38-430mm) |
Still not finding the right product?
Explore all of our products under Nalgene® media bottles, style 2020
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?