Zmień widok
| Rozmiar/SKU | Dostępność | Cena netto |
|---|---|---|
10 ea | Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności | 196,00 zł |
Informacje o tej pozycji
NACRES:
NB.72
UNSPSC Code:
41103806
Material:
PTFE-coated , cylindrical
Manufacturer/tradename:
BRAND 137132
196,00 zł
Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności
Pomoc techniczna
Pozwól nam pomócmaterial
PTFE-coated , cylindrical
manufacturer/tradename
BRAND 137132
L
45 mm
bar diam.
8 mm
General description
BRAND® magnetic cylindrical stirring bars are expertly designed for optimal mixing performance, featuring a smooth, round shape that enhances stirring efficiency. Their Alnico V magnetic cores are protected by a robust PTFE (polytetrafluoroethylene) coating, ensuring durability and resistance to chemical damage. These stirring bars are particularly effective for use in large volumes or thick mixtures, making them ideal for a variety of laboratory applications.
Features and Benefits
- BRAND® magnetic cylindrical stirring bars are round with a smooth surface.
- The Alnico V magnetic cores are encapsulated with high-grade, crack-resistant PTFE (polytetrafluoroethylene) coating.
- Ideal for large volumes or highly viscous mixtures.
Other Notes
BR137132 (India)
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description L 45 mm, bar diam. 8 mm | description L 2.5 mm, bar diam. 2.5 mm | description L 3.5 mm, bar diam. 3.5 mm | description L 50 mm, bar diam. 8 mm |
| manufacturer/tradename BRAND 137132 | manufacturer/tradename BRAND 137100 | manufacturer/tradename BRAND, BRAND 137101 | manufacturer/tradename BRAND 137135 |
| material PTFE-coated , cylindrical | material cylindrical , PTFE-coated | material PTFE-coated , cylindrical | material PTFE-coated , cylindrical |
| bar diam. 8 mm | bar diam. 2.5 mm | bar diam. 3.5 mm | bar diam. 8 mm |
| L 45 mm | L 2.5 mm | L 3.5 mm | L 50 mm |
Still not finding the right product?
Explore all of our products under BRAND® magnetic stirring bar, PTFE, cylindrical
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?