Przejdź do
Z328758
BRAND® magnetic stirring bar, PTFE, cylindrical
L 2.5 mm, bar diam. 2.5 mm
Synonim(y):
cylindrical stir bar, magnetic bar, magnetic stir bar, spinbar, stir bar
Wybierz wielkość
Zmień widok
Rozmiar/SKUDostępnośćCena netto
10 ea
Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności
178,00 zł
Informacje o tej pozycji
UNSPSC Code:
41103806
NACRES:
NB.72
eCl@ss:
32100100
Material:
PTFE-coated , cylindrical
Manufacturer/tradename:
BRAND 137100, BRAND
178,00 zł
Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności
​
Pomoc techniczna
Pozwól nam pomóc
Quality Segment
material
PTFE-coated , cylindrical
manufacturer/tradename
BRAND 137100, BRAND
L
2.5 mm
bar diam.
2.5 mm
General description
BRAND® magnetic cylindrical stirring bars are expertly designed for optimal mixing performance, featuring a smooth, round shape that enhances stirring efficiency. Their Alnico V magnetic cores are protected by a robust PTFE (polytetrafluoroethylene) coating, ensuring durability and resistance to chemical damage. These stirring bars are particularly effective for use in large volumes or thick mixtures, making them ideal for a variety of laboratory applications.
Features and Benefits
  • BRAND® magnetic cylindrical stirring bars are round with a smooth surface.
  • The Alnico V magnetic cores are encapsulated with high-grade, crack-resistant PTFE (polytetrafluoroethylene) coating.
  • Ideal for large volumes or highly viscous mixtures.
Other Notes
BR137100 (India)
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Porównaj podobne pozycje
Wyświetl pełne porównanie
Pokaż różnice
1 of 1
Ta pozycja
Z328863Z328782Z328898
description
L 2.5 mm, bar diam. 2.5 mm
description
L 6 mm, bar diam. 3 mm
description
L 3.5 mm, bar diam. 3.5 mm
description
L 7 mm, bar diam. 2 mm
manufacturer/tradename
BRAND, BRAND 137100
manufacturer/tradename
BRAND, BRAND 137103
manufacturer/tradename
BRAND, BRAND 137101
manufacturer/tradename
BRAND 137106
material
PTFE-coated , cylindrical
material
cylindrical , PTFE-coated
material
PTFE-coated , cylindrical
material
PTFE-coated , cylindrical
L
2.5 mm
L
6 mm
L
3.5 mm
L
7 mm
bar diam.
2.5 mm
bar diam.
3 mm
bar diam.
3.5 mm
bar diam.
2 mm
Quality Level
200
Quality Level
-
Quality Level
200
Quality Level
200
Still not finding the right product?
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)