Wybierz wielkość
| Rozmiar/SKU | Dostępność | Cena netto |
|---|---|---|
6 ea | Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności | 143,00 zł |
Informacje o tej pozycji
143,00 zł
material
AR-Glas®
Quality Segment
description
total delivery Ex + 15 sec waiting time
feature
class AS
packaging
pack of 6 ea
manufacturer/tradename
BRAND 29705
capacity
3 mL
General description
According to DIN EN ISO 648.
Other Notes
Legal Information
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description capacity 3 mL, one mark | description capacity 6 mL, one mark | description capacity 5 mL, one mark | description capacity 2.5 mL, one mark |
| capacity 3 mL | capacity 6 mL | capacity 5 mL | capacity 2.5 mL |
| feature class AS | feature class AS | feature class AS | feature class AS |
| manufacturer/tradename BRAND 29705 | manufacturer/tradename BRAND 29708 | manufacturer/tradename BRAND 29707 | manufacturer/tradename BRAND 29704 |
| material AR-Glas® | material AR-Glas® | material AR-Glas® | material AR-Glas® |
| packaging pack of 6 ea | packaging pack of 6 ea | packaging pack of 6 ea | packaging pack of 6 ea |
| description total delivery Ex + 15 sec waiting time | description total delivery Ex + 15 sec waiting time | description total delivery Ex + 15 sec waiting time | description total delivery Ex + 15 sec waiting time |
Still not finding the right product?
Explore all of our products under BRAND® BLAUBRAND® bulb pipette, calibrated to deliver (TD, EX)
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?