Merck
Z324973
BRAND® BLAUBRAND® bulb pipette, calibrated to deliver (TD, EX)
capacity 6 mL, one mark
Synonim(y):
BLAUBRAND® Pipette, blue-scale bulb pipette, class AS pipette, glass pipette, precision bulb pipette
Informacje o tej pozycji
NACRES:
NB.43
UNSPSC Code:
41121500
Capacity:
6 mL
Material:
AR-Glas®
Feature:
class AS
Packaging:
pack of 6 ea
Pomoc techniczna
Pozwól nam pomóc
material
AR-Glas®
Quality Segment
description
total delivery Ex + 15 sec waiting time
feature
class AS
packaging
pack of 6 ea
manufacturer/tradename
BRAND 29708
capacity
6 mL
General description
Additional to DIN
All BLAUBRAND bulb pipettes are supplied with a batch number and an accompanying batch certificate in the original packaging.
According to DIN EN ISO 648.
Other Notes
BR29708 (India)
Legal Information
AR-Glas is a registered trademark of Schott AG
BLAUBRAND is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
Still not finding the right product?
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)