Wybierz wielkość
| Do Państwa/SKU | Dostępność | Cena netto |
|---|---|---|
1 ea | Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności | 149,00 zł |
Informacje o tej pozycji
149,00 zł
material
amber soda-lime glass , round bottle
joint
joint 29/32 standard ground joint, joint ST/NS neck29/32
feature
without cap
packaging
pack of 1 ea
manufacturer/tradename
BRAND 127065
capacity
2 L
diam. × H
160 mm × 200 mm
General description
Other Notes
Legal Information
1 of 1
Ta pozycja | |||
|---|---|---|---|
| manufacturer/tradename BRAND 127065 | manufacturer/tradename BRAND 126965 | manufacturer/tradename BRAND 23320 | manufacturer/tradename BRAND 129060 |
| feature without cap | feature without cap | feature without cap | feature cap |
| capacity 2 L | capacity 2 L | capacity 2 L | capacity 1000 mL |
| material amber soda-lime glass , round bottle | material clear soda-lime glass , round bottle | material amber borosilicate glass 3.3 , round bottle | material low-density polyethylene , narrow-mouth bottle, round bottle, translucent bottle |
| joint joint ST/NS neck29/32, joint 29/32 standard ground joint | joint joint 29/32 standard ground joint, joint ST/NS neck29/32 | joint joint 29/32 standard ground joint, joint ST/NS neck29/32 | joint - |
| packaging pack of 1 ea | packaging pack of 1 ea | packaging pack of 1 ea | packaging - |
Still not finding the right product?
Explore all of our products under BRAND® reservoir bottle for automatic burettes
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?