Z290777
Właściwości
47 mm
connector for stopper
Opis
Pokaż różnice
1 of 1
Sigma-Aldrich
Vacuum filtration assembly replacement parts
Z290564
Z290416
Sigma-Aldrich® vacuum filtration assembly
Z290505
PTFE gasket
aluminum clamp for 47mm assemblies
for 47 mm filters with glass support, NS 40/35 joints, 4 L flask (included)
for 47 mm filters with stainless steel screen, #8 stopper, same as Z290432 but without side arm flask
-
Explore all of our products under Vacuum filtration assembly replacement parts
Wybierz jedną z najnowszych wersji:
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Dokumenty section.
Odwiedź Bibliotekę dokumentów
Powiązane kategorie
Cell Culture & Cryogenics
Cell Culture Supplies
Filtration
Labware
Vacuum Filters